Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

1,5-Pentanedithiol ≥98%

Flammable
Irritant
Supplier:   Ambeed
1,5-Pentanedithiol ≥98%
Supplier:  Ambeed
Certificates
1,5-Pentanedithiol ≥98%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

 : for Research Use Only

Synonyms:  1,5-Dimercaptopentane
  • CAS Number:928-98-3
  • Formula:C₅H₁₂S₂
  • MDL Number:MFCD00004908
  • Molecular Weight:136.28 g/mol
  • UN:3336
  • ADR:3,Ⅲ
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $19.00
    $19.00
    Each(1/Itm/Bx)
       $19.00

    Supplier Number: A304396-5G
    Specifications:
    New Product
    New Product
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $67.00
    $67.00
    Each(1/Itm/Bx)
       $67.00

    Supplier Number: A304396-25G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.