Ethyl hex-5-ynoate ≥98%
About this item
Caution: for Research Use Only
- CAS Number:108545-38-6
- Formula:C₈H₁₂O₂
- MDL Number:MFCD22023921
- Molecular Weight:140.179 g/mol
- UN:3272
- ADR:3,Ⅲ
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Caution: for Research Use Only
- CAS Number:108545-38-6
- Formula:C₈H₁₂O₂
- MDL Number:MFCD22023921
- Molecular Weight:140.179 g/mol
- UN:3272
- ADR:3,Ⅲ
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: A1159699-100MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A1159699-250MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A1159699-1GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



