Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
PCR Racks

PCR Racks

Supplier:   NEST SCIENTIFIC
PCR Racks
Supplier:  NEST SCIENTIFIC
Certificates

2 Options Available Below

Product Details & Documents

Thes PCR racks fills all Nest PCR products.

  • Stackable
  • Autoclavable
Product Family Options

Product Information

  • Color
    No. of Holes
    For use with
    Material
    Packaging
    Availability
    Price
  • Blur rack, transparent lid
    96
    PCR tubes
    PP
    1 rack and 1 lid, 1/pack, 25/case
    Log in for availability
    Log in for availability
    Case of 25
    Case of 25 - $102.12
    $102.12
    Case of 25
       $102.12

    Supplier Number: 407001
  • Blur rack, transparent lid
    96
    PCR tubes
    PP
    5 racks and 1 lid, 5/pack, 25/case
    Log in for availability
    Log in for availability
    Case of 25
    Case of 25 - $85.40
    $85.40
    Case of 25
       $85.40

    Supplier Number: 407101
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.