Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Malt Extract Agar

Supplier:   RPI
Malt Extract Agar
Supplier:  RPI
Certificates
Malt Extract Agar

5 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Malt Extract Agar is used for the isolation, cultivation and maintenance of yeasts and molds. Formulation is similar to Malt Extract Broth with the addition of agar and glycerol. This broth medium was recommended by Thom and Church for the cultivation of fungi. Its low acidity inhibits the growth of most bacteria. The high carbohydrate content enhances the rapid growth of fungi and yeasts.

 : Research or further manufacturing use only, not for food or drug use.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 76346-776
  • 76346-772
  • 76346-774
  • 77510-316
  • 77510-314
  • CAS No.:
  • 8002-48-0
  • 8002-48-0
  • 8002-48-0
  • 8002-48-0
  • 8002-48-0
  • Solubility:
  • Water with heat
  • Water with heat
  • Water with heat
  • Water with heat
  • Water with heat

Specifications

Product Family Options

Product Information

  • Size
    pH
    Pack type
    Storage temperature
    Availability
    Price
  • 500 g
    At 25 °C: 4.6 ±0.2
    Plastic bottle
    RT
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $177.98
    $177.98
    Each (500g)
       $177.98

    Supplier Number: M21500-500.0
    Specifications:
    More
    Specifications
    Cat. No.
    76346-776
    CAS No.
    8002-48-0
    Solubility
    Water with heat
  • 1 kg
    At 25 °C: 4.6 ±0.2
    Plastic bottle
    RT
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $291.30
    $291.30
    Each (1kg)
       $291.30

    Supplier Number: M21500-1000.0
    Specifications:
    More
    Specifications
    Cat. No.
    76346-772
    CAS No.
    8002-48-0
    Solubility
    Water with heat
  • 2 kg
    At 25 °C: 4.6 ±0.2
    Plastic bottle
    RT
    Log in for availability
    Log in for availability
    Each (2kg)
    Each (2kg) - $552.14
    $552.14
    Each (2kg)
       $552.14

    Supplier Number: M21500-2000.0
    Specifications:
    More
    Specifications
    Cat. No.
    76346-774
    CAS No.
    8002-48-0
    Solubility
    Water with heat
  • 5 kg
    At 25 °C: 4.6 ±0.2
    Plastic bottle
    RT
    Log in for availability
    Log in for availability
    Each
    Each - $1,141.62
    $1,141.62
    Each
       $1,141.62

    Supplier Number: M21500-5000.0
    Specifications:
    More
    Specifications
    Cat. No.
    77510-316
    CAS No.
    8002-48-0
    Solubility
    Water with heat
  • 10 kg
    At 25 °C: 4.6 ±0.2
    Plastic bottle
    RT
    Log in for availability
    Log in for availability
    Each
    Each - $2,139.44
    $2,139.44
    Each
       $2,139.44

    Supplier Number: M21500-10000.0
    Specifications:
    More
    Specifications
    Cat. No.
    77510-314
    CAS No.
    8002-48-0
    Solubility
    Water with heat

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.