Centrifuge Tubes, 15 and 50 ml
Supplier: NEST SCIENTIFIC
Certificates
About this item
Centrifuge tubes could be commonly used to collect cells and separate biomacromolecules by centrifugation in biochemistry and other biological fields. They can also be used to store photosensitive samples and special chemiluminescence reagent.
- Printed graduations (with marking area)
- Polypropylene tube
- High centrifugal strength up to 12000×g
- Flat markable cap with leakproof seal
- One-hand operation design for the caps provides easy handling
- Sterilized by E-beam, SAL=10⁻⁶
- Non-pyrogenic, DNase/Rnase free
Premium level tubes meets the requirements of analytical level, high strength non-toxic materials and raw material comply with USP VI requirements.
Standard level tubes made from virgin polypropylene to meet the general requirements of the laboratory, skirted and self-standing.