Deep Well Plates
About this item
The deep well plates have a variety of specifications, conforming to the needs of fully automatic workstations and experiments.
- High quality imported PP material employed for high stability and no chemical reactions with test reagents
- Maximum sustainable centrifuge force 4000×g
- Compatible with DMSO and inert to water
- Can be stored under subzero temperature from −40 to −80 ℃
- Autoclavable at 20 psi, 121 ℃ for 20 minutes, great heating uniformity
- Minimum residual liquid, low heavy metal content
Genomic DNA extraction, plasmid DNA extraction, and nucleic acid extraction and purification of various samples. High-throughput automatic liquid operation to achieve high-throughput operations, such as protein precipitation, liquid extraction, and processing of animal tissues, bacteria, plants, soil, clinical samples, yeasts and more.
These plates have flat bottom and can be transported for long distance if sealed by heat sealable film. These are produced and packed in Class 100000 clean workshop.
They have alphabetical sorting and corner cut marking for convenient tracking of samples. Each package has a separate article number/batch number identification, facilitating the quality tracking.
Certifications: Certified DNase/RNase and pyrogen free. Conformed to international SBS standards.
39 Options Available Below
Product Details & Documents
The deep well plates have a variety of specifications, conforming to the needs of fully automatic workstations and experiments.
- High quality imported PP material employed for high stability and no chemical reactions with test reagents
- Maximum sustainable centrifuge force 4000×g
- Compatible with DMSO and inert to water
- Can be stored under subzero temperature from −40 to −80 ℃
- Autoclavable at 20 psi, 121 ℃ for 20 minutes, great heating uniformity
- Minimum residual liquid, low heavy metal content
Genomic DNA extraction, plasmid DNA extraction, and nucleic acid extraction and purification of various samples. High-throughput automatic liquid operation to achieve high-throughput operations, such as protein precipitation, liquid extraction, and processing of animal tissues, bacteria, plants, soil, clinical samples, yeasts and more.
These plates have flat bottom and can be transported for long distance if sealed by heat sealable film. These are produced and packed in Class 100000 clean workshop.
They have alphabetical sorting and corner cut marking for convenient tracking of samples. Each package has a separate article number/batch number identification, facilitating the quality tracking.
Certifications: Certified DNase/RNase and pyrogen free. Conformed to international SBS standards.
Product Family Options
Round well
Product Information
- VolumeSterilityWell bottom shapePackagingAvailabilityPrice
- 77508-142Deep well plate, 48-wellDeep well plate, 48-well3.5 mlNon sterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$244.02Supplier Number: 504102Specifications:
More
Specifications
Cat. No.77508-142 - 77508-144Deep well plate, 48-wellDeep well plate, 48-well3.5 mlSterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$268.70Supplier Number: 504162Specifications:
More
Specifications
Cat. No.77508-144 - 77508-096Deep well plate, 96-wellDeep well plate, 96-well0.4 mlNon sterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$207.46Supplier Number: 501102Specifications:
More
Specifications
Cat. No.77508-096 - 77508-098Deep well plate, 96-wellDeep well plate, 96-well0.4 mlSterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$232.14Supplier Number: 501162Specifications:
More
Specifications
Cat. No.77508-098 - 77508-094Deep well plate, 96-wellDeep well plate, 96-well0.5 mlNon sterileV-bottom10/pack, 50/caseLog in for availabilityCase of 50$208.79Supplier Number: 501101Specifications:
More
Specifications
Cat. No.77508-094 - 77508-100Deep well plate, 96-wellDeep well plate, 96-well0.5 mlSterileV-bottom10/pack, 50/caseLog in for availabilityCase of 50$233.57Supplier Number: 501601Specifications:
More
Specifications
Cat. No.77508-100 - 77508-106Deep well plate, 96-wellDeep well plate, 96-well1.0 mlNon sterileV-bottom5/pack, 50/caseLog in for availabilityCase of 50$208.79Supplier Number: 502101Specifications:
More
Specifications
Cat. No.77508-106 - 77508-108Deep well plate, 96-wellDeep well plate, 96-well1.0 mlNon sterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$198.25Supplier Number: 502102Specifications:
More
Specifications
Cat. No.77508-108 - 77508-110Deep well plate, 96-wellDeep well plate, 96-well1.0 mlSterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$223.03Supplier Number: 502162Specifications:
More
Specifications
Cat. No.77508-110 - 77508-112Deep well plate, 96-wellDeep well plate, 96-well1.0 mlSterileV-bottom5/pack, 50/caseLog in for availabilityCase of 50$233.57Supplier Number: 502601Specifications:
More
Specifications
Cat. No.77508-112 - 77508-194Deep well plate, 96-wellDeep well plate, 96-well1.3 mlNon sterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$176.84Supplier Number: 507001Specifications:
More
Specifications
Cat. No.77508-194 - 77508-196Deep well plate, 96-wellDeep well plate, 96-well1.3 mlSterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$176.84Supplier Number: 507101Specifications:
More
Specifications
Cat. No.77508-196 - 77508-122Deep well plate, 96-wellDeep well plate, 96-well2.0 mlNon sterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$230.91Supplier Number: 503102Specifications:
More
Specifications
Cat. No.77508-122 - 77508-124Deep well plate, 96-wellDeep well plate, 96-well2.0 mlSterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$255.69Supplier Number: 503162Specifications:
More
Specifications
Cat. No.77508-124 - 77508-084Deep well plate, 96-wellDeep well plate, 96-well0.36 mlNon sterileV-bottom5/pack, 50/caseLog in for availabilityCase of 50$207.46Supplier Number: 500101Specifications:
More
Specifications
Cat. No.77508-084 - 77508-086Deep well plate, 96-wellDeep well plate, 96-well0.36 mlSterileV-bottom5/pack, 50/caseLog in for availabilityCase of 50$232.14Supplier Number: 500161Specifications:
More
Specifications
Cat. No.77508-086
Square well
Product Information
- VolumeSterilityWell bottom shapePackagingAvailabilityPrice
- 77508-152Deep well plate, 24-wellDeep well plate, 24-well10.4 mlNon sterileV-bottom5/pack, 50/caseLog in for availabilityCase of 50$248.01Supplier Number: 504301Specifications:
More
Specifications
Cat. No.77508-152 - 77508-154Deep well plate, 24-wellDeep well plate, 24-well10.4 mlSterileV-bottom5/pack, 50/caseLog in for availabilityCase of 50$271.67Supplier Number: 504361Specifications:
More
Specifications
Cat. No.77508-154 - 77508-204Deep well plate, 24-wellDeep well plate, 24-well16 mlSterileV-bottom1/pack, 25/caseLog in for availabilityCase of 25$117.86Supplier Number: 510001Specifications:
More
Specifications
Cat. No.77508-204 - 77508-138Deep well plate, 48-wellDeep well plate, 48-well4.6 mlNon sterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$213.81Supplier Number: 504002Specifications:
More
Specifications
Cat. No.77508-138 - 77508-140Deep well plate, 48-wellDeep well plate, 48-well4.6 mlSterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$238.49Supplier Number: 504062Specifications:
More
Specifications
Cat. No.77508-140 - 77508-082Deep well plate, 96-wellDeep well plate, 96-well0.5 mlNon sterileV-conical bottom5/pack, 50 /caseLog in for availabilityCase of 50$259.99Supplier Number: 500021Specifications:
More
Specifications
Cat. No.77508-082 - 77508-088Deep well plate, 96-wellDeep well plate, 96-well0.5 mlSterileV-conical bottom5/pack, 50/caseLog in for availabilityCase of 50$274.33Supplier Number: 500621Specifications:
More
Specifications
Cat. No.77508-088 - 77508-090Deep well plate, 96-wellDeep well plate, 96-well1.0 mlNon sterileU-bottom5/pack, 50 /caseLog in for availabilityCase of 50$196.40Supplier Number: 501002Specifications:
More
Specifications
Cat. No.77508-090 - 77508-092Deep well plate, 96-wellDeep well plate, 96-well1.0 mlSterileU-bottom5/pack, 50 /caseLog in for availabilityCase of 50$221.18Supplier Number: 501062Specifications:
More
Specifications
Cat. No.77508-092 - 77508-102Deep well plate, 96-wellDeep well plate, 96-well1.6 mlNon sterileU-bottom5/pack, 50 /caseLog in for availabilityCase of 50$196.40Supplier Number: 502002Specifications:
More
Specifications
Cat. No.77508-102 - 77508-104Deep well plate, 96-wellDeep well plate, 96-well1.6 mlSterileU-bottom5/pack, 50 /caseLog in for availabilityCase of 50$221.18Supplier Number: 502062Specifications:
More
Specifications
Cat. No.77508-104 - 77508-114Deep well plate, 96-wellDeep well plate, 96-well2.2 mlNon sterileV-bottom5/pack, 50 /caseLog in for availabilityCase of 50$208.79Supplier Number: 503001Specifications:
More
Specifications
Cat. No.77508-114 - 77508-116Deep well plate, 96-wellDeep well plate, 96-well2.2 mlNon sterileU-bottom5/pack, 50 /caseLog in for availabilityCase of 50$208.79Supplier Number: 503002Specifications:
More
Specifications
Cat. No.77508-116 - 77508-118Deep well plate, 96-wellDeep well plate, 96-well2.2 mlNon sterileV-conical bottom5/pack, 50 /caseLog in for availabilityCase of 50$279.96Supplier Number: 503201Specifications:
More
Specifications
Cat. No.77508-118 - 77508-120Deep well plate, 96-wellDeep well plate, 96-well2.2 mlSterileU-bottom5/pack, 50/caseLog in for availabilityCase of 50$233.57Supplier Number: 503062Specifications:
More
Specifications
Cat. No.77508-120 - 77508-130Deep well plate, 96-wellDeep well plate, 96-well2.2 mlSterileV-bottom5/pack, 50 /caseLog in for availabilityCase of 50$233.57Supplier Number: 503501Specifications:
More
Specifications
Cat. No.77508-130 - 77508-132Deep well plate, 96-wellDeep well plate, 96-well2.2 mlSterileV-conical bottom5/pack, 50/caseLog in for availabilityCase of 50$303.92Supplier Number: 503261Specifications:
More
Specifications
Cat. No.77508-132 - 77508-134Deep well plate, 96-wellDeep well plate, 96-well2.2 mlNon sterileI-shaped, U-bottom5/pack, 50/caseLog in for availabilityCase of 50$118.78Supplier Number: 503711Specifications:
More
Specifications
Cat. No.77508-134 - 77508-136Deep well plate, 96-wellDeep well plate, 96-well2.2 mlSterileI-shaped, U-bottom5/pack, 50/caseLog in for availabilityCase of 50$118.78Supplier Number: 503761Specifications:
More
Specifications
Cat. No.77508-136 - 77508-156Deep well plate, 384-wellDeep well plate, 384-well120 µlNon sterileV-bottom5/pack, 50/caseLog in for availabilityCase of 50$288.36Supplier Number: 504701Specifications:
More
Specifications
Cat. No.77508-156 - 77508-158Deep well plate, 384-wellDeep well plate, 384-well240 µlNon sterileV-bottom5/pack, 50/caseLog in for availabilityCase of 50$288.36Supplier Number: 504703Specifications:
More
Specifications
Cat. No.77508-158 - 77508-160Deep well plate, 384-wellDeep well plate, 384-well120 µlSterileV-bottom5/pack, 50/caseLog in for availabilityCase of 50$306.12Supplier Number: 504761Specifications:
More
Specifications
Cat. No.77508-160 - 77508-162Deep well plate, 384-wellDeep well plate, 384-well240 µlSterileV-bottom5/pack, 50/caseLog in for availabilityCase of 50$309.86Supplier Number: 504763Specifications:
More
Specifications
Cat. No.77508-162
Silicone sealing mats
Product Information
- Catalog #Temperature rangeDescriptionPackagingSterilityAvailabilityPrice
- −50...+200 °CSilicone sealing mat, round well, 96-well, 2 ml10/pack, 50/caseNon sterileLog in for availabilityCase of 50$233.27
Plastic comb
Product Information
- Catalog #DescriptionAvailabilityPrice
- Plastic comb, 24 tip, sterile, compatible with Thermo KingFisher System, 1/pack, 25/caseLog in for availabilityCase of 25$161.48