Orders LinkContactUsLinkComponent
Pasteur Pipettes
Supplier:   NEST SCIENTIFIC
Pasteur Pipettes
Supplier:  NEST SCIENTIFIC
Certificates
Pasteur Pipettes
9 Options Available Below
Product Details & Documents
Pipette can be twisted for use with micro vessels and special-shaped containers.
  • Good chemical stability
  • Transparent with clear graduations for easy observation
  • High precision and good repeatability
  • Individually wrapped, sterilized by EO
  • DNase/RNase, endotoxin and pyrogenic free
Product Family Options
Product Information
  • Material
    Volume
    Length
    Sterility
    Packed
    Availability
    Price
  • 77507-580
    Pasteur pipette
    Pasteur pipette
    PP
    1 ml
    144 mm
    Sterile
    2000/case
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $226.61
    $226.61
    Case of 2,000
       $226.61
    Supplier Number: 318012
  • 77507-582
    Pasteur pipette
    Pasteur pipette
    PP
    1ml
    144 mm
    Non sterile
    5000/case
    Log in for availability
    Log in for availability
    Case of 5,000
    Case of 5,000 - $252.93
    $252.93
    Case of 5,000
       $252.93
    Supplier Number: 318031
  • 77507-584
    Pasteur pipette
    Pasteur pipette
    PP
    2 ml
    150 mm
    Sterile
    2000/case
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $226.61
    $226.61
    Case of 2,000
       $226.61
    Supplier Number: 318112
  • 77507-586
    Pasteur pipette
    Pasteur pipette
    PP
    2 ml
    150 mm
    Non sterile
    5000/case
    Log in for availability
    Log in for availability
    Case of 5,000
    Case of 5,000 - $252.93
    $252.93
    Case of 5,000
       $252.93
    Supplier Number: 318131
  • 77507-588
    Pasteur pipette
    Pasteur pipette
    PP
    3 ml
    162 mm
    Sterile
    2000/case
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $226.61
    $226.61
    Case of 2,000
       $226.61
    Supplier Number: 318212
  • 77507-590
    Pasteur pipette
    Pasteur pipette
    PP
    3 ml
    162 mm
    Non sterile
    5000/case
    Log in for availability
    Log in for availability
    Case of 5,000
    Case of 5,000 - $252.93
    $252.93
    Case of 5,000
       $252.93
    Supplier Number: 318231
  • 77507-592
    Pasteur pipette, extra long
    Pasteur pipette, extra long
    PP
    3 ml
    182 mm
    Sterile
    2000/case
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $261.94
    $261.94
    Case of 2,000
       $261.94
    Supplier Number: 318314
  • 77507-596
    Pasteur pipettes
    Pasteur pipettes
    PP
    5 ml
    Sterile
    250/pack, 1000/case
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $230.20
    $230.20
    Case of 1,000
       $230.20
    Supplier Number: 318516
  • 77507-594
    Pasteur pipettes
    Pasteur pipettes
    PP
    10 ml
    Sterile
    125/pack, 500/case
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $262.41
    $262.41
    Case of 500
       $262.41
    Supplier Number: 318417