Standard stirrers are designed for general-purpose laboratory use.
- Microprocessor controlled
- Enhanced electronic features
- Chemical-resistant housing
- Now with Best-in-Class, Exclusive 5-Year Warranty
These analog units are microprocessor controlled and are equipped with easy-to-use, dial-adjustment controls
Stirrers are available with ceramic or aluminum top plates. Ceramic top plates feature an easy-to-clean, chemical-resistant, reflective white surface. Aluminum top plates will not crack or chip. The unit's housing is constructed of a polymer that is chemical-resistant.
Enhanced electronics regulate stirring functions. A ramping feature gradually increases speed to prevent splashing, improve magnetic coupling, and provide excellent low end control. Speed is precisely controlled, with consistent stirring at all speeds powered by a dependable, continuous-duty motor. A low-profile design requires minimal bench space and permits use within a fume hood. The spill-resistant housing channels fluids away from internal components in the event of a spill.