About this item
Ice free cool boxes with ISO 9001 and 13485 certified, ice free cool boxes are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
- Free of ice core and modules
- various refrigeration sources and modules are available
Cold core: (Precooling to 0.5 - 4 ℃) Put in minus 20 ℃ more than 4 hours, at indoor temperature placed 10 minutes before use. (Indoor temperature) Put in minus 20 ℃ more than 4 hours, direct use, put in minus 80 ℃ more than 4 hours, direct use.
Frozen core: (Precooling in −20 ℃ half an hour) Put in minus 20 ℃ more than 6 hours, Put in minus 80 ℃ more than 3 hours.
Cold Core:
−20 ℃ Ultra cold storage freezer temperature: 4 hours+ freezing time
−80 ℃ Ultra cold storage freezer temperature: 2 hours+ freezing time
Freeze Core:
−20 ℃ Ultra cold storage freezer temperature: 6 hours+ freezing time
−80 ℃ Ultra cold storage freezer temperature: 3 hours+ freezing time