VWR® Cork Sheet
3 mm (⅛"), 101 x 305 mm (4 x 12")
23420-708
Use to protect workbenchtop from heat, acids, and alkalies.
- Protects against heat, acids, and alkalies
Cork sheet can also be used as a dissecting pad.
- Dimensions:101 x 305 mm (4 x 12")
- Thickness:3 mm (⅛")
- Cat. No.:23420-708
- Environmentally Preferable:Low Manufacturing Impact



