ContactUsLinkComponent
Stainless Steel Indicator for Industial Applications, Defender 6000
Stainless Steel Indicator for Industial Applications, Defender 6000
Catalog # 77435-062
Supplier:  Ohaus
undefined
Stainless Steel Indicator for Industial Applications, Defender 6000
Catalog # 77435-062
Supplier:  Ohaus
Supplier Number:  30575584
Specifications
  • Model:
    INDICATOR DT61XWE
  • Description:
    Stainless steel indicator for industial applications, Defender 6000 series
  • Environmentally Preferable:
  • Cat. No.:
    77435-062
Specifications
About this item
Washdown bench stainless steel indicator for standard industrial applications. Defender 6000 indicator, constructed from 316 stainless steel, stands up to harsh cleaning in processing and packaging environments with IP 68 immersion/IP 69K washdown protection.
  • White LED displays clear results for fast processing
  • The indicator's configurable color bar matrix allows for detailed package weight checking to ensure compliance
  • Knife-proof numeric touch keypad, indicator is suitable in harsh processing environments
The Defender 6000 Series extreme washdown indicator is specifically designed for food, chemical and pharmaceutical applications including production, processing, and packaging. Defender 6000 handles the most demanding applications with a 316 stainless steel housing equipped with bar matrix check light, knife-proof front panel and EasyConnect™.