- Depth:24"
- Description:Cupboard, 1 shelf, 1 left door
- Height:34½"
- Length:24"
- Material:Plastic laminate
- Cat. No.:33501-084
- Supplier no.:B0110362424
Case systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.
- Plastic laminate provides superior resistance to chemicals, scratching, and abrasion
- The high-quality laboratory grade plastic laminate cabinets are manufactured to exact tolerances with consistent quality standards
- The unique hardware system provides rigid support for vertical integration without limiting adjustability
The key component of case systems casework is the plastic laminate. A perfect choice for laboratory environments. The radiused 3 mm PVC edging on doors and drawers further protects the casework from dents and marring while eliminating sharp edges and corners.
Construction features an industrial-grade, environmentally responsible WOODSTALK® engineered board core from Dow BioProducts, using formaldehyde-free resin to bind wheat-straw fiber, and heavy-duty institutional grade hardware. The result cabinets that are as attractive as they are durable.
Case cabinetry gives you the adaptability necessary in today’s laboratories. Vertical supports are slotted, allowing infinite height adjustment of overhead cabinets, electrical raceways, services, and reagent shelves.
Cupboard cabinet has one adjustable shelf and one door, which swings left. Standing height base cabinets are common for traditional laboratory and technical environments. They are usually installed for permanent or semi-permanent layouts. Standing height base cabinets are 24" deep and 34.5" high for standing applications. Color: Beige.