Order Entry
Puerto Rico
ContactUsLinkComponent
Casework, Laminate, Sitting Height Base Cabinets, Cupboard and Drawer Cabinet
Casework, Laminate, Sitting Height Base Cabinets, Cupboard and Drawer Cabinet
Catalog # 33501-138
Supplier:  Case Systems
Small Business Enterprise
undefined
Casework, Laminate, Sitting Height Base Cabinets, Cupboard and Drawer Cabinet
Catalog # 33501-138
Supplier:  Case Systems
Supplier Number:  B3110302424
Small Business Enterprise

Specifications

  • Depth:
    24"
  • Description:
    Cabinet with 1 shelf, 1 left door, 1 drawer
  • Height:
    29½"
  • Length:
    24"
  • Material:
    Plastic laminate
  • Cat. No.:
    33501-138
  • Supplier no.:
    B3110302424

Specifications

About this item

Case systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.

  • The key component of case systems casework is the plastic laminate
  • Cupboard cabinet has one shelf, one door, and one drawer

A perfect choice for laboratory environments, plastic laminate provides superior resistance to chemicals, scratching, and abrasion. The radiused 3 mm PVC edging on doors and drawers further protects the casework from dents and marring while eliminating sharp edges and corners.

The high-quality laboratory grade plastic laminate cabinets are manufactured to exact tolerances with consistent quality standards. Construction features an industrial-grade, environmentally responsible WOODSTALK® engineered board core from Dow BioProducts, using formaldehyde-free resin to bind wheat-straw fiber, and heavy-duty institutional grade hardware. The result - cabinets that are as attractive as they are durable.

Case cabinetry gives the adaptability necessary in today's laboratories. The unique hardware system provides rigid support for vertical integration without limiting adjustability. Vertical supports are slotted, allowing infinite height adjustment of overhead cabinets, electrical raceways, services, and reagent shelves.

Door swings left. Sitting height base cabinets are common for traditional laboratory and technical environments. They are usually installed for permanent or semi-permanent layouts. Color: Beige.

Ordering information: Case Systems laminate laboratory furniture has all the components necessary for a complete laboratory set-up. For further information, contact your Avantor Furniture Specialist.