Centrifuge, Low Noise and Fast Lifting Speed
Supplier: NEST SCIENTIFIC
Certificates
About this item
Safe and convenient, low noise and fast lifting speed electronic locks. Centrifugation only when the lid is locked. The lid opens automatically after centrifugation, low noise inside the cavity.
- Easy to use and easy to observe
- Capacitive and responsive touchpad
- LCD display with real-time display of speed, time and operating status
- Centrifugal time and speed adjustable
Suitable for 0.2, 0.5, 1.5, 5 ml centrifuge tubes without change the rotor.