ContactUsLinkComponent
ABS Plastic Indicator for Industial Applications, Defender 3000
Catalog # 77435-132
Supplier:  Ohaus
undefined
ABS Plastic Indicator for Industial Applications, Defender 3000
Catalog # 77435-132
Supplier:  Ohaus
Supplier Number:  30659310
Specifications
  • Model:
    INDICATOR DT33P
  • Description:
    ABS plastic indicator for industial applications, Defender 3000
  • Environmentally Preferable:
  • Cat. No.:
    77435-132
Specifications
About this item
ABS plastic indicator for basic industrial applications.
  • Results are clearly shown using the large, bright backlit LCD display
  • In Checkweighing mode, the display changes color (yellow/green/red) to alert when it is within the target weight range
Designed with affordability in mind for basic industrial applications, the i-DT33P indicator combines high-impact ABS housing with a large multicolor backlit LCD display and bi-directional RS232. The i-DT33P also features software for Weighing, Parts Counting, Checkweighing, and Totalization.