Universal Pipette Tips
About this item
Nest Scientific Universal Fit Pipette Tips are made from premium-grade 100% polypropylene and engineered for the most demanding molecular biology applications.
Universal fit: Suitable for use with most pipettors
- Withstands organic solvents
- Less residue
- Made from low retention plastic
Sterilized by gamma irradiation (only valid for some variants), they are certified RNase-, DNase-, human DNA-, and pyrogen-free.
: Available in different packaging options: Bulk, racked, stack racked or in refills.
24 Options Available Below
Product Details & Documents
Nest Scientific Universal Fit Pipette Tips are made from premium-grade 100% polypropylene and engineered for the most demanding molecular biology applications.
Universal fit: Suitable for use with most pipettors
- Withstands organic solvents
- Less residue
- Made from low retention plastic
Sterilized by gamma irradiation (only valid for some variants), they are certified RNase-, DNase-, human DNA-, and pyrogen-free.
: Available in different packaging options: Bulk, racked, stack racked or in refills.
Product Family Options
Bulk, non sterile
Product Information
- Tip styleVolumeSterileTip colorPackedAvailabilityPrice
- Supplier Number: 301001
- Supplier Number: 304001
- Supplier Number: 302001
- Supplier Number: 305001
- Supplier Number: 303001
- Supplier Number: 306001
Racked, sterile
Product Information
- Tip styleVolumeSterileTip colorPackedAvailabilityPrice
- Supplier Number: 301012
- Supplier Number: 304012
- Supplier Number: 302012
- Supplier Number: 305012
- Supplier Number: 303012
- Supplier Number: 306012
Refills, non sterile
Product Information
- Tip styleVolumeSterileTip colorPackedAvailabilityPrice
- Supplier Number: 301023
- Graduated, extra long10 µl-Clear96/rack, 960/pack, 9600 caseLog in for availabilityCase of 9,600$334.25Supplier Number: 304023
- Supplier Number: 302023
- Supplier Number: 305023
- Graduated1000 µl (1 ml)-Clear96/rack, 960/pack, 9600 caseLog in for availabilityCase of 4,800$199.58Supplier Number: 303023
- Graduated1250 µl (1,25 ml)-Clear96/rack, 960/pack, 9600 caseLog in for availabilityCase of 4,800$199.68Supplier Number: 306023
Double stack racked, sterile
Product Information
- Tip styleVolumeSterileTip colorPackedAvailabilityPrice
- Supplier Number: 301052
- Graduated, extra long10 µl+Clear96/wafer, 960/pack, 9600 caseLog in for availabilityCase of 9,600$342.12Supplier Number: 304052
- Supplier Number: 302052
- Supplier Number: 305052
Bagged, non sterile
Product Information