Order Entry
United States
ContactUsLinkComponent
Homogenizer, Bead Ruptor 24
Homogenizer, Bead Ruptor 24
  77547-556
New Product
undefined
Homogenizer, Bead Ruptor 24
  77547-556
 :  19-070
New Product

 

  • Description:
    Homogenizer, Bead Ruptor 24
  • Environmentally Preferable:
  • Processing Range:
    25 µl - 2 ml
  • Electrical:
    100 - 230 VAC, 50/60 Hz
  • No. of cycles:
    1 - 10
  • Size range:
    0.8 - 6.5 m/s
  • Weight:
    29 kg (65 lbs.)
  • W×D×H:
    38.1×43.2×33 cm (15×17×13")
  • Cat. No.:
    77547-556

 

 

The bead ruptor 24 homogenizer is ideal for demanding laboratories seeking unmatched reliability, power and efficiency in grinding, lysing and homogenization of even the toughest samples.

  • Fixed 2 ml tube carriage - process 24×2 ml tubes
  • Convenient front loading design
  • Programmable memory settings
  • Optimized tube motion ensures the highest performance
  • Ideal for: DNA/RNA extraction, tissue homogenization, enzyme isolation and protein purificaion

The bead ruptor 24 bead mill homogenizer is designed with a unique carriage motion to ensure that the intra-tube bead movement reduces swirling and results in the highest bead impact forces of any bead mill on the market. The increased power of the bead ruptor 24 decreases processing time and reduces sample heating to maximize sample preparation efficiency.

 : TUV, CE and UKCA approved