Cleanroom Hazard Warning Tape, Ultratape
11212-060
- Width:5.1 cm (2")
- Color:Black/Yellow
- Length:16.5 m (18 yds.)
- Cat. No.:11212-060
- Supplier no.:1165BY200P2D
Use these conformable, vinyl black and yellow striped hazard warning tapes to designate the OSHA critical areas of cleanrooms.
: Clean and double-bagged. Also available in red and white striped and green and white striped; contact your Avantor sales representative.