HelixMark® PharmaFocus® Silicone Tubing
Supplier: Freudenberg Medical
Certificates
About this item
Designed specifically to fit the needs of the pharmaceutical marketplace.
- Fully autoclavable and sterilizable
- Ideal for use in transfer operations as well as other applications requiring non-leachable material
- Material certification in every package
- Lot traceability available
- Length: 50' coils.
All tubing provides exceptional biocompatibility, having undergone studies according to ISO 10993-1 and meeting USP Class VI and FDA 21 CFR 177.2600 requirements. Manufactured under Good Manufacturing Practices (GMP) in an FDA-registered and ISO 13485 compliant medical device facility.
The platinum-cured silicone elastomer used for PharmaFocus® tubing undergoes a battery of biocompatibility tests appropriate for pharmaceutical applications.
These processes eliminate costly extended implant studies typically performed on biomedical grade silicones, resulting in an economical silicone tubing that delivers consistent quality performance.
Ordering information: Packaged double-bagged in crush-resistant boxes. Material certification and lot traceability included in every box.