PowerHead™ Foam Mops, Micronova
Supplier: Micronova
Certificates
About this item
Disposable mop heads are offered in a variety of densities and materials
Polyester-covered foam mop heads 68400-346 are available both nonsterile and sterile for critical cleaning. Wide-pore foam mop head 68400-340 is used for cleanroom support and maintenance areas.
Stainless steel PowerHead™ handles 68400-312 and -314 hold and wring the various mop heads. Autoclavable handles are 71.1cm (28") long. A quick-disconnect fitting on the handle allows the user to customize length with single-piece and extendable universal handles (see 68400-302 series).
Mop heads and handles are available in 25.4cm (10") and 35.6cm (14") widths.
Ordering information: All components are ordered separately.