Hypersil GOLD™ PEI HILIC HPLC Column
Catalog # 76477-176
Supplier: Thermo Scientific
Specifications
- Column Format:Preparative
- Ø int.:10 mm
- Particle Size:5 µm
- Pore size:175 Å
- Length:250 mm
- Endcapped:No
- pH Range:2 - 8
- Carbon Load:6%
- Cat. No.:76477-176
- Packing material:Polyethylenimin (PEI)
Specifications
About this item
Highly polar and hydrophilic analytes that are difficult to analyze with conventional C18 columns can be retained with Thermo Scientific™ Hypersil GOLD™ PEI HILIC LC columns for hydrophilic interaction chromatography.
These columns enable straightforward separations without sample derivatization or ion-pairing reagents. The columns feature excellent peak shape and selectivity and offer improved sensitivity in MS detection.
With particle sizes of 1.9, 3 and 5 µm, Hypersil GOLD PEI-HILIC columns are ideal for applications in drug development as well as in food and environmental analysis.