Order Entry
Puerto Rico
ContactUsLinkComponent
BioBasic™ PREP SEC 120 Size Exclusion Chromatography HPLC Column
BioBasic™ PREP SEC 120 Size Exclusion Chromatography HPLC Column
Catalog # 76420-878
Supplier:  Thermo Scientific
BioBasic™ PREP SEC 120 Size Exclusion Chromatography HPLC Column
Catalog # 76420-878
Supplier:  Thermo Scientific
Supplier Number:  73405-307846A

Specifications

  • Ø int.:
    7.8 mm
  • Particle Size:
    5 µm
  • Pore size:
    120 Å
  • Max. Pressure:
    4300 psi
  • Length:
    300 mm
  • Endcapped:
    No
  • pH Range:
    2 - 8
  • Surface Area:
    300 m²/g
  • Temperature Limit:
    60 °C
  • Carbon Load:
    >8.2%
  • Cat. No.:
    76420-878
  • Packing material:
    Spherical, fully porous, ultrapure silica

Specifications

About this item

BioBasic™ HPLC column provide outstanding size separation of water soluble compounds.

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases

BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 0.7 to 120 kilodaltons, allowing for the analysis of proteins, peptides and other biomolecules.