Order Entry
United States
ContactUsLinkComponent
MINISTAR 80 control Electronic Overhead Stirrers
MINISTAR 80 control Electronic Overhead Stirrers
Catalog # 76329-868
Supplier:  IKA Works
undefined
MINISTAR 80 control Electronic Overhead Stirrers
Catalog # 76329-868
Supplier:  IKA Works
Supplier Number:  0025005082

Specifications

  • Description:
    MINISTAR 80 control overhead stirrer
  • Environmentally Preferable:
  • Cat. No.:
    76329-868

Specifications

About this item

This overhead stirrer is ideal for special applications. Combining high performance with excellence, the units require minimum space.

  • Enclosed in hardened glass
  • Fast, response display for maximum visibility and chemical resistance
  • Clear display shows all essential information at a glance
  • Viscosities up to 60000 mPas and volumes of up to 50000 ml
  • Speed is continuously adjustable between 0/30 - 500 rpm
Certifications: IP protection class according to DIN EN 60529: IP 54.
Ordering information: Supplied without accessories, a wide range of accessories are available to order separately.