Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Calcium chloride 2M in aqueous solution

Calcium chloride 2M in aqueous solution

Irritant
Supplier:   RPI
Calcium chloride 2M in aqueous solution
Supplier:  RPI
Certificates
Calcium chloride 2M in aqueous solution

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Caution: Research or further manufacturing use only, not for food or drug use.

Synonyms:  Chloro calcium
  • CAS Number:10035-04-8
  • Formula:CaCl₂·2H₂O
  • MDL Number:MFCD00149613
  • Molecular Weight:147.01 g/mol
  • Boiling Pt:100 °C (212 °F)
  • Melting Pt:0°C ( 32°F)
  • Density:1.086 g/ml at 20 °C
  • Storage Temperature:Room Temperature
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear Solution
  • Concentration:
    2M
  • Sterility:
    Pass Test
  • BSE/TSE Free:
    BSE/TSE Free
  • Country of Origin:
    US
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    50 ml
    Log in for availability
    Log in for availability
    Each (50ml)
    Each (50ml) - $53.75
    $53.75
    Each (50ml)
       $53.75

    Supplier Number: C25100-50.0
  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    Each
    Each - $94.33
    $94.33
    Each
       $94.33

    Supplier Number: C25100-1000.0

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.