Order Entry
Puerto Rico
ContactUsLinkComponent
SPILFYTER® TIDYMAT™ Mat Roll, NPS
SPILFYTER® TIDYMAT™ Mat Roll, NPS
Catalog # 56609-227
Supplier:  NPS
undefined
SPILFYTER® TIDYMAT™ Mat Roll, NPS
Catalog # 56609-227
Supplier:  NPS
Supplier Number:  521048

Specifications

  • Dimensions:
    45.7 cm x 45.7 m (18" x 150')
  • Cat. No.:
    56609-227
  • Supplier no.:
    521048

Specifications

About this item

Mat absorbs non-aggressive spills and prevents slipping

Use mat on walkways in plant and office areas, as shelf and drawer liners, and around machinery. Can be cut to fit. Mat is made of recycled fiber and stands up to heavy traffic. Absorbs up to nine times its own weight.