Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Spermidine trihydrochloride
Spermidine trihydrochloride ≥99%, Calbiochem®
Spermidine trihydrochloride ≥99%, Calbiochem®
Spermidine trihydrochloride ≥99%, Calbiochem®
80058-662
:
MilliporeSigma
:
Spermidine trihydrochloride ≥99%, Calbiochem®
80058-662
:
MilliporeSigma
:
56766-5GM
:
Pack type:
Bottle
Size:
5 g
Cat. No.:
80058-662
Supplier no.:
56766-5GM
:
:
254.63 g/mol
:
12,08893
:
256…258 °C
:
Freezer
:
206-379-0
:
334-50-9
:
C₇H₂₂Cl₃N₃
:
MFCD00012918