ContactUsLinkComponent
Pierce™ Inclusion Body Solubilization Reagent, Thermo Scientific
Pierce™ Inclusion Body Solubilization Reagent, Thermo Scientific
Catalog # PI78115
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Inclusion Body Solubilization Reagent, Thermo Scientific
Catalog # PI78115
Supplier:  Invitrogen
Supplier Number:  78115
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Inclusion Body Solubilization Reagent
  • Cat. No.:
    PI78115
  • Volume:
    100 ml
  • Supplier no.:
    78115
Specifications
About this item
Inclusion Body Solubilization Reagent effectively retrieves recombinant proteins that form insoluble aggregates during bacterial overexpression.
  • Achieves the essential first step for subsequent insoluble protein refolding procedures
  • One easy to use solution
  • Compatible with Coomassie Plus Protein Assay Kit to determine degree of expression
  • Proprietary denaturant can be easily removed by dialysis so that SDS-PAGE can be performed
  • Can accommodate additional components such as reducing agents or chelating agents