Phenolic Foam Spill Control Pillows, NPS
Supplier: Avantor
Certificates
About this item
Convenient pillows contain and absorb hazardous and toxic spills quickly
- Able to absorb 98% of their capacity in 30 seconds
- Pillows absorb a total of 10 to 20 times their weight of almost any liquid
- Treat through elution, evaporation, or incineration in accordance with local regulations
Spill control pillows feature a nonpolluting, chemically inert glass foam to absorb acids, caustics, and organic solvents.
Caution: Not intended for use with strong oxidizers.
Frequently Bought Together