Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Imidazole

Imidazole

Corrosive
Irritant
Health
Supplier:   RPI
Imidazole
Supplier:  RPI
Certificates
Imidazole

6 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

 : Research or further manufacturing use only, not for food or drug use.

Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Storage Temperature:Room Temperature
  • EINECS:206-019-2
  • UN:3263
  • ADR:8,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off white crystals
  • Assay:
    ≥ 99.0 %
  • Identification:
    Pass Test
  • Insolubles:
    ≤ 0.1 %
  • Loss on Drying:
    ≤ 0.5 %
  • Melting Point:
    87.0 to 91.0 oC
  • OPTICAL @ 260 NM:
    ≤ 0.2
  • pH (5%, water) @25 oC:
    9.5 to 10.5
  • Solubility (20%, water):
    Pass
  • DNase:
    Not Detected
  • Protease:
    Not Detected
  • RNase:
    Not Detected
  • BSE/TSE Free:
    BSE/TSE Free
  • Country of Origin:
    Report
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 10 g
    Log in for availability
    Log in for availability
    Each (10g)
    Each (10g) - $39.13
    $39.13
    Each (10g)
       $39.13

    Supplier Number: I52000-10.0
  • 50 g
    Log in for availability
    Log in for availability
    Each
    Each - $114.82
    $114.82
    Each
       $114.82

    Supplier Number: I52000-50.0
  • 100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $155.08
    $155.08
    Each (100g)
       $155.08

    Supplier Number: I52000-100.0
  • 500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $424.89
    $424.89
    Each (500g)
       $424.89

    Supplier Number: I52000-500.0
  • 1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $699.07
    $699.07
    Each (1kg)
       $699.07

    Supplier Number: I52000-1000.0
  • 5 kg
    Log in for availability
    Log in for availability
    Each (5kg)
    Each (5kg) - $3,049.59
    $3,049.59
    Each (5kg)
       $3,049.59

    Supplier Number: I52000-5000.0

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.