Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Formamide

Formamide

Health
Supplier:   RPI
Formamide
Supplier:  RPI
Certificates
Formamide

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For nucleic acid hybridization and sequencing.

Caution: Research or further manufacturing use only, not for food or drug use.

Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:200-842-0
Specifications for Product Family

Specification Test Results

  • Appearance:
    Colorless liquid
  • Assay:
    ≥ 99.5 %
  • Freezing Point:
    1.0 - 3.0 °C
  • Copper:
    ≤ 0.0001 %
  • Iron:
    ≤ 0.0005 %
  • Lead (Pb):
    ≤ 0.0001 %
  • Zinc:
    ≤ 0.0001 %
  • Country of Origin:
    DE
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $28.21
    $28.21
    Each (100ml)
       $28.21

    Supplier Number: F11900-0.1
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $111.69
    $111.69
    Each (500ml)
       $111.69

    Supplier Number: F11900-0.5
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 1 L
    Log in for availability
    Log in for availability
    Each
    Each - $179.04
    $179.04
    Each
       $179.04

    Supplier Number: F11900-1000.0
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.