Calbiochem® ALLM
Catalog # 80055-990
Supplier: MilliporeSigma
CAS Number:
Specifications
- Size:25 mg
- Protein/Peptide Name:ALLM
- Cat. No.:80055-990
- Supplier no.:208721-25MG
Specifications
About this item
Inhibitor of calpain I (Ki=120nM), calpain II (Ki=230nM), cathepsin B (Ki=100nM), and cathepsin L (Ki=0.6nM). Inhibits activation-induced programmed cell death and restores defective immune responses in HIV+ donors. Also prevents nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene.