Order Entry
United States
ContactUsLinkComponent
Thrombin
Thrombin
  95017-586
 :  Cytiva
Product available on GSA Advantage®
 :  
undefined
Thrombin
  95017-586
 :  Cytiva
 :  27-0846-01
Product available on GSA Advantage®
 :  

 

  • CAS No.:
    9002-04-4
  • Source:
  • Species:
  • Size:
    500 units
  • Environmentally Preferable:
  • Form:
    White solid
  • Reconstitution Instructions:
    Cleavage buffer: PBS: 140 mM NaCl, 2.7 mM KCl, 10 mM Na₂HPO₄, 1.8 mM KH₂PO₄, pH 7.3
    Elution buffer: 50 mM Tris-HCl, 10 mM reduced glutathione, pH 8.0
  • Enzyme Name:
    Thrombin
  • Molecular Weight:
    37000 [Mr]
  • Tested Applications:
    Site-specific separation of the GST tag from proteins expressed using pGEX-T vectors (pGEX-4T-1, pGEX-4T-2, pGEX-4T-3, pGEX-2TK)
  • Cat. No.:
    95017-586
  • Supplier no.:
    27-0846-01

 

 

Thrombin is a protease used to digest fusion proteins prepared from pGEX vectors containing the recognition sequence for thrombin (pGEX-1lT, pGEX-2T, pGEX-2TK, pGEX-4T-1, pGEX-4T-2, and pGEX-4T-3).

  • Enables the site-specific cleavage of fusion proteins containing an accessible thrombin recognition sequence
  • Purified from bovine plasma; functionally free of other clotting factors, plasminogen, and plasmin
  • Can be used either following affinity purification or while fusion proteins are bound to Glutathione Sepharose™ 4B, Glutathione Sepharose™ High Performance, or Glutathione Sepharose™ 4 Fast Flow

Application: Used for site-specific separation of the GST tag from proteins expressed using pGEX-T vectors.

Cleavage Buffer: PBS 140 mM NaCl, 2,7 mM KCl, 10 mM Na2HPO4, 1,8 mM KH2PO4, pH 7.3
Elution Buffer:50 mM Tris-HCl, 10 mM reduced glutathione, pH 8.0

Inhibitors: Pefabloc TH benzamidine, AEBSF, APMSF, antithrombin III, Antipain, α1-antitrypsin, aprotinin, chymostatin, hirudin, leupeptin, PMSF

Molecular Weight: 37000

Unit Definition: One unit of enzyme cleaves 90% of 100 μg of a test GST fusion protein when incubated in 1X PBS at 22 °C for 16 hours

 : Supplied as lyophilized powder. Includes 50 mM Tris-HCl, 10 mM reduced glutathione, pH 8.0.