Verapamil hydrochloride ≥99% (by perchloric acid titration), Calbiochem®, Sigma-Aldrich®
80055-494
:
- Pack type:Aluminum drum
- Size:100 mg
- Cat. No.:80055-494
- Supplier no.:676777-100MG
:
:491.07 g/mol
:Ambient
:152-11-4
:23313-68-0
:C₂₇H₃₈N₂O₄
:MFCD00055208