Ellagic acid dihydrate ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80058-054
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:500 mg
- Cat. No.:80058-054
- Supplier no.:324683-500MG
Specifications
About this item
:
:302.19 g/mol
:M-Clarity™ quality level = MQ100
EINECS:207-508-3
:GHS07
Storage Temperature:Ambient
CAS Number:133039-73-3
:476-66-4
Formula:C₁₄H₆O₈
MDL Number:MFCD00006914