Pierce™ Western Blotting Kits, Far-Western Blot Kit for Biotinylated Proteins, Thermo Scientific
PI23500
:
- Description:Far-Western Blot Kit for Biotinylated Proteins, for 10 Mini Gel or Blot Experiments
- Cat. No.:PI23500
- Supplier no.:23500
The Pierce™ Far-Western Blot Kit for Biotinylated Proteins provide a nonisotopic approach to far-Western analysis that can be performed either directly in the gel or on the membrane
- Reliable and sensitive detection method offers a practical and safe alternative to radiolabeling the bait protein
- Primary antibody-free detection - uses a biotinylated protein as the probe, eliminating the need for primary antibodies or even to have prior knowledge of target-protein identities
- Compatible with both SDS-PAGE and native gels
- Biotin/streptavidin-HRP systems demonstrate less nonspecific binding compared to antibodies
- Gels can be used for total protein staining after the in-gel chemiluminescent detection step, eliminating the need to run two gels
On-membrane detection offers greater sensitivity whereas in-gel detection is faster and prevents problems associated with incomplete or inefficient transfer.
: Kit includes Streptavidin-HRP Conjugate (0.1 mg), 10X Dilution Buffer (50 mL), PBS Pack (makes 500 mL, 17 pack), Luminol Enhancer (55 mL), Stable Peroxide Buffer (55 mL), Surfact-Amps 20 Detergent Solution (6x10 mL) and Pre-cut Cellophane (10 sheets).