ContactUsLinkComponent
Static Free™ Hand Guard® Lotion, ITW Chemtronics
Static Free™ Hand Guard® Lotion, ITW Chemtronics
Catalog # 15145-450
Supplier:  ITW Chemtronics
undefined
Static Free™ Hand Guard® Lotion, ITW Chemtronics
Catalog # 15145-450
Supplier:  ITW Chemtronics
Supplier Number:  C805
Specifications
  • Capacity:
    237 ml (8 oz.)
  • Description:
    Pump Bottle
  • Cat. No.:
    15145-450
  • Supplier no.:
    C805
Specifications
About this item
This hand lotion is formulated to protect against electrostatic charge
Nongreasy formula contains aloe vera and Vitamin E. Contains no lanolin, silicone, or mineral oil that could inhibit solderability of printed circuits or components. The emollients penetrate deeply to moisten and refresh dry, chapped, or chemically irritated skin.