About this item
Unique shape and style provide excellent eye protection in a variety of frames and lenses.
- Wide viewing area provides good visibility and protection from flying particles
- Nylon frame and lens with molded-in side shields and brow guard
- Four-stop temple adjustment (130 to 150 mm) provides a face-hugging fit
- Optically correct, one-piece polycarbonate lens is lightweight and impact-resistant
- Lens angle adjusts for a comfortable fit
- Temples will not scratch the lens when folded
Models with Duoflex™ temples have soft, behind-the-ear comfort cushions. Astrospec 3000 Slim glasses are 10 mm smaller to fit slender facial profiles. Astrospec 3000 Custom Bridge models are designed to fit low-profile nasal areas and high-profile cheeks. Select Ultra-dura® lens coating for scratch and UV protection, or Uvextreme® antifog coating. To replace worn, damaged, or scratched lenses, just snap replacement lenses into frame; no tools are required.
Clear lenses are for most indoor applications. Gray and silver mirror lenses minimize eye fatigue due to outdoor sunlight and glare. Amber lenses are useful for low-light applications where enhanced contrast is needed. SCT-Low IR lenses are ideal for when peripheral IR protection is required. The Patriot® RWB frame is a red, white, and blue design with miniature U.S. flags at temples and “United We Stand” inscribed on bottom right lens outside field of vision.