ContactUsLinkComponent
Synthware Distillation Head, Hickman-Hinkle, With Side Port, Microscale, Kemtech America
  60004-142
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Distillation Head, Hickman-Hinkle, With Side Port, Microscale, Kemtech America
  60004-142
 :  H141410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
  • Cat. No.:
    60004-142
  • [ST] joint size:
    14/10
  • Supplier no.:
    H141410
Hickman-Hinkle still head with threaded side port for syringe access.
  • Heavy wall borosilicate construction
  • Threaded upper joint
  • Open cap included
  • Hickman-Hinkle syle still head
For microscale fractional distillation. Supplied with a nylon compression cap, Viton* O-ring, and side port cap with 8 mm PTFE-lined silicone septum.