ContactUsLinkComponent
Synthware Hickman Distillation Head, Microscale, Kemtech America
Synthware Hickman Distillation Head, Microscale, Kemtech America
Catalog # 60004-134
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Hickman Distillation Head, Microscale, Kemtech America
Catalog # 60004-134
Supplier:  Kemtech America
Supplier Number:  H101410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Cat. No.:
    60004-134
  • [ST] joint size:
    14/10
  • Supplier no.:
    H101410
Specifications
About this item
Hickman still head for microscale fractional distillation
  • Heavy wall borosilicate construction
  • Threaded upper joint
  • Open cap included
  • Hickman style still head
Supplied with a nylon compression cap and Viton* O-ring.