ContactUsLinkComponent
Synthware Adapter, Vacuum Takeoff, Threaded, Microscale, Kemtech America
Synthware Adapter, Vacuum Takeoff, Threaded, Microscale, Kemtech America
Catalog # 60004-114
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Adapter, Vacuum Takeoff, Threaded, Microscale, Kemtech America
Catalog # 60004-114
Supplier:  Kemtech America
Supplier Number:  A621410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • [ST] joint:
    14/10
  • Cat. No.:
    60004-114
  • Supplier no.:
    A621410
Specifications
About this item
This is a vacuum takeoff joint adapter suitable for microscale applications.
  • Threaded
  • Nylon compression cap and Viton® O-ring included
Delivery: Supplied with a nylon compression cap and Viton® O-ring.