ContactUsLinkComponent
Synthware Adapter, Distillation, Threaded, Microscale, Kemtech America
Synthware Adapter, Distillation, Threaded, Microscale, Kemtech America
Catalog # 60004-110
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Adapter, Distillation, Threaded, Microscale, Kemtech America
Catalog # 60004-110
Supplier:  Kemtech America
Supplier Number:  A351410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • [ST] joint:
    14/10
  • Cat. No.:
    60004-110
  • Supplier no.:
    A351410
Specifications
About this item
Distillation joint adapter is suitable for microscale applications.
  • Threaded
  • For microscale applications
  • Compression cap and O-ring included
Threaded distillation joint adapter is supplied with a nylon compression cap and Viton® O-ring.