ContactUsLinkComponent
Synthware Adapter, Thermometer, with Hose Connection, Microscale, Kemtech America
Synthware Adapter, Thermometer, with Hose Connection, Microscale, Kemtech America
  60004-106
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Adapter, Thermometer, with Hose Connection, Microscale, Kemtech America
  60004-106
 :  A781410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
  • [ST] joint:
    14/10
  • Cat. No.:
    60004-106
  • Supplier no.:
    A781410
Vacuum joint adapter with vertically adjustable condenser tube for microscale applications.
  • Vacuum-tight seal
  • Cap accommodates thermometer
  • Nylon compression cap and Viton® O-ring included
  • With hose connection
Supplied with a nylon compression cap and Viton® O-ring to form a vacuum tight seal. To use as a syringe port, replace the O-ring with PTFE -lined septum. Cap accommodates a 5.0 to 7.5 mm (ext. Ø) thermometer.