ContactUsLinkComponent
Synthware Adapter, Inlet or Thermometer, Microscale, Kemtech America
Synthware Adapter, Inlet or Thermometer, Microscale, Kemtech America
Catalog # 60004-104
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Adapter, Inlet or Thermometer, Microscale, Kemtech America
Catalog # 60004-104
Supplier:  Kemtech America
Supplier Number:  A771410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • [ST] joint:
    14/10
  • Cat. No.:
    60004-104
  • Supplier no.:
    A771410
Specifications
About this item
This is a joint adapter suitable for microscale applications.
  • Vacuum-tight seal
  • Cap accommodates thermometer
  • Nylon compression cap and Viton® O-ring included
Supplied with a nylon compression cap and Viton® O-ring to form a vacuum tight seal. To use as a syringe port, replace the O-ring with PTFE -lined septum. Cap accommodates a 5.0 to 7.5 mm ext. Ø thermometer.