ContactUsLinkComponent
VWR® Premium Formalin Neutralizing Pads
VWR® Premium Formalin Neutralizing Pads
Catalog # 89131-874
Supplier:  VWR International
undefined
VWR® Premium Formalin Neutralizing Pads
Catalog # 89131-874
Supplier:  VWR International
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Dimensions:
    40.6W x 50.8L (16 x 20")
  • Cat. No.:
    89131-874
Specifications
About this item
Durable, tear-resistant pads are manufactured with a bonded chemical that helps to minimize exposure to formaldehyde vapors.
  • Offensive Formalin Odors Eliminated
  • Disposable
  • Durable
  • Tear-Resistant
  • Blue Surface Provides Contrast
Pads immediately absorb and neutralize excess formalin run-off during dissection
They are also useful for lining shelves or transport containers where leakage may occur.
The distinctive blue surface of the pads offers an excellent contrast to even small-tissue specimens.