ContactUsLinkComponent
UltraSOLV® ScrubTIPS® Cleaning Swabs with Grit Tips, Foamtec International
UltraSOLV® ScrubTIPS® Cleaning Swabs with Grit Tips, Foamtec International
Catalog # 10809-942
undefined
UltraSOLV® ScrubTIPS® Cleaning Swabs with Grit Tips, Foamtec International
Catalog # 10809-942
Supplier Number:  HT170060-5
Specifications
  • Description:
    Silicon carbide grit tip, 600-Grit
  • Length:
    15.2 cm (6")
  • Cat. No.:
    10809-942
  • Tip style:
    Rigid
  • Supplier no.:
    HT170060-5
Specifications
About this item
These unique cleaning tools are ideal for polishing stains, scratches, and process residue from hard-to-clean surfaces such as electro-static chucks
Swabs are available with a dual tip or rigid tip and washed diamond or silicon carbide abrasives.
Ideal for removing flashing from sensitive medical devices and optics and maintaining high-precision injection molds.