Synthware Stopper with #15 O-Ring Joint, Kemtech America
Catalog # 60004-852
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:#15 O-Ring Joint Stopper
- Environmentally Preferable:
- Cat. No.:60004-852
- Supplier no.:S480015
Specifications
About this item
Designed for use with manifolds equipped with #15 O-rings.
- Heavy wall borosilicate construction
- Ground glass joints
- #15 O-ring
Supplied complete with one #116 Viton* O-ring.