Specifications
- Description:Compresstome VF-210-0Z and VF-510-0Z blade holder
- Material:Stainless steel, magnetic
- Cat. No.:77435-416
Specifications
About this item
Replacement blade holder for Compresstome vibrating microtome.
- Each blade holder securely holds ceramic and/or steel blades
- Blade holder attaches to vibrating head, and is easily removable
For use on the VF-210-0Z or VF-510-0Z Compresstome vibratome, the magnetic blade holder features magnets for easy attachment of stainless steel blades. It is compatable with ceramic blades as well.