Specifications
- Description:Oxygenation attachment
- Cat. No.:77435-530
Specifications
About this item
This is an oxygenation tube compatible with compresstome models, used to assist in the process of keeping samples alive longer.
- Clips onto the buffer tank to help buffer solutions stay oxygenated during the cutting process
- Used for experiments involving fresh/acute/live tissue slices that require oxygen
Oxygenation attachment used to attach to the buffer tray of the compresstome tissue slicers.