ContactUsLinkComponent
Compresstome® Magnetic Blade Holder for VF-210-0Z and VF-510-0Z
Compresstome® Magnetic Blade Holder for VF-210-0Z and VF-510-0Z
Catalog # 77435-420
undefined
Compresstome® Magnetic Blade Holder for VF-210-0Z and VF-510-0Z
Catalog # 77435-420
Supplier Number:  VFBHVMMGNT
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Compresstome VF-210-0Z and VF-510-0Z magnetic blade holder
  • Cat. No.:
    77435-420
Specifications
About this item
This is a magnetic blade holder for easy stainless steel blade installation. This can also be used for ceramic blades.
  • Premium magnetic blade holder for the compresstome vibrating microtome
  • Each blade holder has built-in magnets to securely hold stainless steel blades without glue
  • Ceramic and tungsten blades can also be used with glue
Packaging: Includes one magnetic blade holder, one allen wrench, and one small screw.