ContactUsLinkComponent
Compresstome® Plastic Cleanup Razors
Compresstome® Plastic Cleanup Razors
Catalog # 77435-426
undefined
Compresstome® Plastic Cleanup Razors
Catalog # 77435-426
Supplier Number:  VFBLVMPCR
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Compresstome plastic cleanup razors
  • Cat. No.:
    77435-426
Specifications
About this item
Used to assist in cleaning super glue and other material from the specimen tube.
  • Pack of five plastic razors
Plastic cleanup razor for cleaning the surface of compresstome specimen tubes. Used for cleaning off of the white plunger of specimen tubes after sectioning. Designed to not damage the white plunger.